Phenyl ether, 99%
Phenyl ether, 99%
Phenyl ether, 99%
Thermo Scientific Chemicals
Phenyl ether, 99%
CAS: 101-84-8 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA130605000
Price (MYR)
297.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
297.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenyl ether
CAS101-84-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN