m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
Thermo Scientific Chemicals
m-Phenylenediamine, 99+%
CAS: 108-45-2 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA130565000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialm-Phenylenediamine
CAS108-45-2
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0