L-Phenylalanine, 98.5+%
L-Phenylalanine, 98.5+%
L-Phenylalanine, 98.5+%
L-Phenylalanine, 98.5+%
Thermo Scientific Chemicals
L-Phenylalanine, 98.5+%
CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA130315000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Phenylalanine
Melting Point270.0°C to 275.0°C
CAS63-91-2
pH5.4 to 6.0 (1% aq. soln.)