Phenylacetyl chloride, 98%
Phenylacetyl chloride, 98%
Phenylacetyl chloride, 98%
Phenylacetyl chloride, 98%
Thermo Scientific Chemicals
Phenylacetyl chloride, 98%
CAS: 103-80-0 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA130305000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenylacetyl chloride
Name Note98%
CAS103-80-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement