2-Phenoxyethanol, 99%
2-Phenoxyethanol, 99%
2-Phenoxyethanol, 99%
2-Phenoxyethanol, 99%
Thermo Scientific Chemicals
2-Phenoxyethanol, 99%
2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C | CAS: 122-99-6 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number FSA130242500
Price (MYR)
231.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
231.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Phenoxyethanol
CAS122-99-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0