Phenothiazine, 99%
Phenothiazine, 99%
Phenothiazine, 99%
Thermo Scientific Chemicals
Phenothiazine, 99%
CAS: 92-84-2 | C12H9NS | 199.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number FSA130210050
Price (MYR)
1,971.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,971.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenothiazine
CAS92-84-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
If s