Phenethyl alcohol, 99%
Phenethyl alcohol, 99%
Phenethyl alcohol, 99%
Phenethyl alcohol, 99%
Phenethyl alcohol, 99%
CAS: 60-12-8 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA130182500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenethyl alcohol
CAS60-12-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED