Phenazine methosulfate, 98%
Phenazine methosulfate, 98%
Phenazine methosulfate, 98%
Phenazine methosulfate, 98%
Thermo Scientific Chemicals
Phenazine methosulfate, 98%
CAS: 299-11-6 | C13H11N2·CH3O4S | 306.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
10 g
Packaging:
Glass bottle
Catalog number FSA130160250
Price (MYR)
3,547.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,547.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenazine methosulfate
Name Note99%
CAS299-11-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement