Phenazine, 98%
Phenazine, 98%
Phenazine, 98%
Thermo Scientific Chemicals
Phenazine, 98%
CAS: 92-82-0 | C12H8N2 | 180.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA130150100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenazine
Name Note98%
CAS92-82-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.