1,10-Phenanthroline Monohydrochloride Monohydrate, 97%
1,10-Phenanthroline Monohydrochloride Monohydrate, 97%
1,10-Phenanthroline Monohydrochloride Monohydrate, 97%
1,10-Phenanthroline Monohydrochloride Monohydrate, 97%
Thermo Scientific Chemicals
1,10-Phenanthroline Monohydrochloride Monohydrate, 97%
CAS: 18851-33-7 | HCl·H2O | 234.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA130141000
Price (MYR)
2,695.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,695.00
EA
Add to cart
Specifications
Chemical Name or Material1, 10-Phenanthroline monohydrochloride monohydrate
CAS18851-33-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0