Phenanthrene, 97%
Phenanthrene, 97%
Phenanthrene, 97%
Thermo Scientific Chemicals
Phenanthrene, 97%
CAS: 85-01-8 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
50 g
Packaging:
View all
Plastic bottle
Catalog number FSA130095000
Price (MYR)
2,306.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,306.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenanthrene
Name Note98+%
CAS85-01-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.