3-Pentanol, 98%
3-Pentanol, 98%
3-Pentanol, 98%
Thermo Scientific Chemicals
3-Pentanol, 98%
CAS: 584-02-1 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA129991000
Price (MYR)
432.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
432.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Pentanol
CAS584-02-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0