DL-2-Pentanol, 98%
DL-2-Pentanol, 98%
DL-2-Pentanol, 98%
Thermo Scientific Chemicals
DL-2-Pentanol, 98%
CAS: 6032-29-7 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA129985000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-2-Pentanol
CAS6032-29-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.