2,4-Pentanedione, 99+%
2,4-Pentanedione, 99+%
2,4-Pentanedione, 99+%
2,4-Pentanedione, 99+%
Thermo Scientific Chemicals
2,4-Pentanedione, 99+%
CAS: 123-54-6 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
2.5 kg
Catalog number FSA129962500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Pentanedione
CAS123-54-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.