Pentaerythritol, 98%
Pentaerythritol, 98%
Pentaerythritol, 98%
Pentaerythritol, 98%
Thermo Scientific Chemicals
Pentaerythritol, 98%
CAS: 115-77-5 | C5H12O4 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA129872500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPentaerythritol
CAS115-77-5
Melting Point257.0°C to 264.0°C
ColorWhite
EINECS Number204-104-9