n-Pentadecanoic acid, 99%
n-Pentadecanoic acid, 99%
n-Pentadecanoic acid, 99%
n-Pentadecanoic acid, 99%
Thermo Scientific Chemicals
n-Pentadecanoic acid, 99%
CAS: 1002-84-2 | C15H30O2 | 242.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA129830050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Pentadecanoic acid
Name Note99%
CAS1002-84-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning