Oxamic acid, 98%
Oxamic acid, 98%
Oxamic acid, 98%
Thermo Scientific Chemicals
Oxamic acid, 98%
CAS: 471-47-6 | C2H3NO3 | 89.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA129621000
Price (MYR)
1,406.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,406.00
EA
Add to cart
Specifications
Chemical Name or MaterialOxamic acid
CAS471-47-6
Health Hazard 1GHS Signal Word: Warning
Melting Point210.0°C