Oxalyl chloride, 98%
Oxalyl chloride, 98%
Oxalyl chloride, 98%
Oxalyl chloride, 98%
Thermo Scientific Chemicals
Oxalyl chloride, 98%
CAS: 79-37-8 | C2Cl2O2 | 126.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA129611000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOxalyl chloride
CAS79-37-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Contact with w
CAS75-44-5
View more
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only