1-Octen-3-ol, 98%
1-Octen-3-ol, 98%
1-Octen-3-ol, 98%
Thermo Scientific Chemicals
1-Octen-3-ol, 98%
CAS: 3391-86-4 | C8H16O | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA129471000
Price (MYR)
845.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
845.00
EA
Add to cart
Specifications
Chemical Name or Material1-Octen-3-ol
CAS3391-86-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0