n-Octane, 99+%, extra pure
n-Octane, 99+%, extra pure
n-Octane, 99+%, extra pure
n-Octane, 99+%, extra pure
Thermo Scientific Chemicals
n-Octane, 99+%, extra pure
CAS: 111-65-9 | C8H18 | 114.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA129375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Octane
CAS111-65-9
CAS Min %97
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0