1-Octadecene, 90%, tech.
1-Octadecene, 90%, tech.
1-Octadecene, 90%, tech.
1-Octadecene, 90%, tech.
Thermo Scientific Chemicals
1-Octadecene, 90%, tech.
CAS: 112-88-9 | C18H36 | 252.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA129310025
Price (MYR)
656.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
656.00
EA
Add to cart
Specifications
Chemical Name or Material1-Octadecene
CAS112-88-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.