Octadecane, 99+%
Octadecane, 99+%
Octadecane, 99+%
Thermo Scientific Chemicals
Octadecane, 99+%
CAS: 593-45-3 | C18H38 | 254.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
Packaging:
Glass bottle
Catalog number FSA129292500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOctadecane
CAS593-45-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.