n-Nonylamine, 98%
n-Nonylamine, 98%
n-Nonylamine, 98%
Thermo Scientific Chemicals
n-Nonylamine, 98%
CAS: 112-20-9 | C9H22ClN | 179.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
View all
Glass Bottle
Catalog number FSA129185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Nonylamine
CAS112-20-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0