Nitroterephthalic acid, 99+%
Nitroterephthalic acid, 99+%
Nitroterephthalic acid, 99+%
Nitroterephthalic acid, 99+%
Thermo Scientific Chemicals
Nitroterephthalic acid, 99+%
CAS: 610-29-7 | C8H5NO6 | 211.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA129000250
Price (MYR)
835.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
835.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNitroterephthalic acid
CAS610-29-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0