4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%
Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, C6H4NNa2O6P.6H2O, CAS Number-333338-18-4 | CAS: 333338-18-4 | C6H4NNa2O6P | 263.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA128860100
Price (MYR)
1,194.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,194.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Nitrophenyl phosphate, disodium salt, hexahydrate
CAS333338-18-4
Melting Point>300.0°C
ColorWhite to Yellow
Infrared SpectrumAuthentic