2-Nitrophenyl-beta-D-galactopyranoside, 99%
2-Nitrophenyl-beta-D-galactopyranoside, 99%
2-Nitrophenyl-beta-D-galactopyranoside, 99%
2-Nitrophenyl-beta-D-galactopyranoside, 99%
Thermo Scientific Chemicals
2-Nitrophenyl-beta-D-galactopyranoside, 99%
CAS: 369-07-3 | C12H15NO8 | 301.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA128820050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Nitrophenyl-β-D-galactopyranoside
CAS369-07-3
CAS Min %98.5
CAS Max %100.0
Melting Point195.0°C