4-Nitrophenylacetic acid, 99%
4-Nitrophenylacetic acid, 99%
4-Nitrophenylacetic acid, 99%
4-Nitrophenylacetic acid, 99%
Thermo Scientific Chemicals
4-Nitrophenylacetic acid, 99%
CAS: 104-03-0 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA128760500
Price (MYR)
479.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
479.00
EA
Add to cart
Specifications
Chemical Name or Material4-Nitrophenylacetic acid
Name Note99%
CAS104-03-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning