4-Nitrophenyl acetate, 97%
4-Nitrophenyl acetate, 97%
4-Nitrophenyl acetate, 97%
4-Nitrophenyl acetate, 97%
Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%
CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number FSA128751000
Price (MYR)
1,087.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,087.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Nitrophenyl acetate
Name Note97%
CAS830-03-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger