5-Nitroisatin, 97%
5-Nitroisatin, 97%
5-Nitroisatin, 97%
Thermo Scientific Chemicals
5-Nitroisatin, 97%
CAS: 611-09-6 | C8H4N2O4 | 192.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA128720050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material5-Nitroisatin
Name Note97%
CAS611-09-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning