4-Nitrobenzoic acid, 99+%
4-Nitrobenzoic acid, 99+%
4-Nitrobenzoic acid, 99+%
4-Nitrobenzoic acid, 99+%
Thermo Scientific Chemicals
4-Nitrobenzoic acid, 99+%
CAS: 62-23-7 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA128461000
Price (MYR)
224.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
224.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Nitrobenzoic acid
Name Note99+%
CAS62-23-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning