3-Nitrobenzaldehyde, 99%
3-Nitrobenzaldehyde, 99%
3-Nitrobenzaldehyde, 99%
3-Nitrobenzaldehyde, 99%
Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%
CAS: 99-61-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA128415000
Price (MYR)
770.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
770.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Nitrobenzaldehyde
Name Note99%
CAS99-61-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning