2-Nitrobenzaldehyde, 99+%
2-Nitrobenzaldehyde, 99+%
2-Nitrobenzaldehyde, 99+%
2-Nitrobenzaldehyde, 99+%
Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%
CAS: 552-89-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA128401000
Price (MYR)
1,020.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,020.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Nitrobenzaldehyde
CAS552-89-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for s