4-Nitroaniline, 99%
4-Nitroaniline, 99%
4-Nitroaniline, 99%
Thermo Scientific Chemicals
4-Nitroaniline, 99%
CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA128375000
Price (MYR)
656.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
656.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Nitroaniline
CAS100-01-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breat
View more
Indicator
RUO – Research Use Only