3-Nitroaniline, 98%
3-Nitroaniline, 98%
3-Nitroaniline, 98%
Thermo Scientific Chemicals
3-Nitroaniline, 98%
CAS: 99-09-2 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA128361000
Price (MYR)
413.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
413.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Nitroaniline
Name Note99+%
CAS99-09-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger