o-Nitroacetophenone, 95%
o-Nitroacetophenone, 95%
o-Nitroacetophenone, 95%
o-Nitroacetophenone, 95%
Thermo Scientific Chemicals
o-Nitroacetophenone, 95%
CAS: 577-59-3 | C8H7NO3 | 165.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number FSA128320025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialo-Nitroacetophenone
Name Note95%
CAS577-59-3
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning