Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
CAS: 75-84-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA128250500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNeopentyl alcohol
CAS75-84-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0