1-Naphthol, 99+%
1-Naphthol, 99+%
1-Naphthol, 99+%
Thermo Scientific Chemicals
1-Naphthol, 99+%
1-Naphthol, 99+%, C10H8O, CAS Number-90-15-3, 1-hydroxynaphthalene, 1-naphthalenol, 1-naphthol, alpha-naphthol, basf ursol ern, fouramine ern, fourrine ern, furro er, naphthol, tertral ern, 100g, 06, 596, 278.0 deg.C to 280.0 deg.C, 100.0, 99.0, CHEBI:10319, Beige-Gray to Brown, 201-969-4, 125 deg.C | CAS: 90-15-3 | C10H8O | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA128195000
Price (MYR)
599.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
599.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Naphthol
Name Note99+%
CAS90-15-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger