Myrcene, 90%, tech., stabilized
Myrcene, 90%, tech., stabilized
Myrcene, 90%, tech., stabilized
Myrcene, 90%, tech., stabilized
Thermo Scientific Chemicals
Myrcene, 90%, tech., stabilized
CAS: 123-35-3 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA128085000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMyrcene
CAS123-35-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
May produce an al
CAS127-91-3