Methylurea, 97%
Methylurea, 97%
Methylurea, 97%
Thermo Scientific Chemicals
Methylurea, 97%
CAS: 598-50-5 | C2H6N2O | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA127995000
Price (MYR)
512.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
512.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethylurea
Name Note97%
CAS598-50-5
CAS Min %96.0
CAS Max %100.0