5-Methyl-DL-tryptophan, 98%
5-Methyl-DL-tryptophan, 98%
5-Methyl-DL-tryptophan, 98%
Thermo Scientific Chemicals
5-Methyl-DL-tryptophan, 98%
CAS: 951-55-3 | C12H14N2O2 | 218.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA127972500
Price (MYR)
680.00
Bottle
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
680.00
Bottle
Specifications
Chemical Name or Material5-Methyl-DL-tryptophan
CAS951-55-3
Melting Point280.0°C to 282.0°C
ColorWhite
EINECS Number213-453-6