Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass Bottle
Catalog number FSA127791000
Price (MYR)
236.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
236.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
Name Note99.7%
CAS67-68-5
Health Hazard 1HYGROSCOPIC