Methyl sulfide, 99+%, extra pure
Methyl sulfide, 99+%, extra pure
Methyl sulfide, 99+%, extra pure
Methyl sulfide, 99+%, extra pure
Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure
CAS: 75-18-3 | C2H6S | 62.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA127760100
Price (MYR)
403.00
BOT
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
403.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfide
CAS75-18-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g