mono-Methyl succinate, 95%
mono-Methyl succinate, 95%
mono-Methyl succinate, 95%
mono-Methyl succinate, 95%
Thermo Scientific Chemicals
mono-Methyl succinate, 95%
CAS: 3878-55-5 | C5H8O4 | 132.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA127730100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialmono-Methyl succinate
Name Note98%
CAS3878-55-5
CAS Min %97.5
CAS Max %100.0