1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, C5H9NO, CAS Number-872-50-4, 1-methyl-2-pyrrolidinone, 1-methyl-2-pyrrolidone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidinone, n-methylpyrrolidone, 2.5L | CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number FSA127630250
Price (MYR)
3,872.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
3,872.00
EA
Add to cart
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24.0°C
CAS872-50-4
CAS Min %98.5
Health Hazard 1Danger
View more
Suitable for battery materials development.
RUO – Research Use Only