1-Methylpyrrolidine, 98%
1-Methylpyrrolidine, 98%
1-Methylpyrrolidine, 98%
1-Methylpyrrolidine, 98%
Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%
CAS: 120-94-5 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA127615000
Price (MYR)
1,409.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,409.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Methylpyrrolidine
CAS120-94-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi