N-Methylpiperidine, 99%
N-Methylpiperidine, 99%
N-Methylpiperidine, 99%
N-Methylpiperidine, 99%
Thermo Scientific Chemicals
N-Methylpiperidine, 99%
CAS: 626-67-5 | C6H14ClN | 135.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA127482500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Methylpiperidine
Name Note99%
CAS626-67-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger