3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
Thermo Scientific Chemicals
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
CAS: 89-25-8 | C10H10N2O | 174.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA127465000
Price (MYR)
577.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
577.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Methyl-1-phenyl-2-pyrazolin-5-one
Name Note99%
CAS89-25-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
View more
Coupling reagent
RUO – Research Use Only