2-Methyl-1-pentene, 99%
2-Methyl-1-pentene, 99%
2-Methyl-1-pentene, 99%
2-Methyl-1-pentene, 99%
Thermo Scientific Chemicals
2-Methyl-1-pentene, 99%
CAS: 763-29-1 | C6H12 | 84.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number FSA127400100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-1-pentene
Name Note99%
CAS763-29-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger