Methyl nicotinate, 99%
Methyl nicotinate, 99%
Methyl nicotinate, 99%
Methyl nicotinate, 99%
Thermo Scientific Chemicals
Methyl nicotinate, 99%
CAS: 93-60-7 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA127215000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl nicotinate
Name Note99%
CAS93-60-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: