2-Methyl-1,4-naphthoquinone, 98%
2-Methyl-1,4-naphthoquinone, 98%
2-Methyl-1,4-naphthoquinone, 98%
2-Methyl-1,4-naphthoquinone, 98%
Thermo Scientific Chemicals
2-Methyl-1,4-naphthoquinone, 98%
CAS: 58-27-5 | C11H8O2 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA127181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-1, 4-naphthoquinone
Name Note98%
CAS58-27-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning